Skip to Content
Merck
All Photos(2)

Key Documents

268879

Sigma-Aldrich

N,N′-Bis(2-hydroxyethyl)ethylenediamine

97%

Synonym(s):

2,2′-(Ethylenediimino)diethanol

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$189.00
50 G
$319.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$189.00
50 G
$319.00

About This Item

Linear Formula:
HOCH2CH2NHCH2CH2NHCH2CH2OH
CAS Number:
Molecular Weight:
148.20
Beilstein:
1739630
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

98-100 °C (lit.)

functional group

amine
hydroxyl

SMILES string

OCCNCCNCCO

InChI

1S/C6H16N2O2/c9-5-3-7-1-2-8-4-6-10/h7-10H,1-6H2

InChI key

GFIWSSUBVYLTRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The ideal-gas enthalpy of formation of N,N′-bis(2-hydroxyethyl)ethylenediamine was studied[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamic properties and ideal-gas enthalpies of formation for dicyclohexyl sulfide, diethylenetriamine, di-n-octyl sulfide, dimethyl carbonate, piperazine, hexachloroprop-1-ene, tetrakis(dimethylamino)ethylene, N, N'-bis-(2-hydroxyethyl) ethylenediamine and 1, 2, 4-triazolo [1, 5-a] pyrimidine.
Steele WV, et al.
Journal of Chemical and Engineering Data, 42(6), 1037-1052 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service