Skip to Content
Merck
All Photos(1)

Documents

268836

Sigma-Aldrich

Perfluorotetracosane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)22CF3
CAS Number:
Molecular Weight:
1238.18
Beilstein:
5474566
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

188-190 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C24F50/c25-1(26,3(29,30)5(33,34)7(37,38)9(41,42)11(45,46)13(49,50)15(53,54)17(57,58)19(61,62)21(65,66)23(69,70)71)2(27,28)4(31,32)6(35,36)8(39,40)10(43,44)12(47,48)14(51,52)16(55,56)18(59,60)20(63,64)22(67,68)24(72,73)74

InChI key

AXFFGMPNSZVEDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The structural change of the evaporated perfluorotetracosane (PFT) films by the mechanical rubbing was studied by X-ray Absorption Near Edge Structure (XANES) spectroscopy.

Application

Perfluorotetracosane was used as a solvent to characterize two high molecular weight fractions of poly(tetrafluoroethylene) (PTFE) by using laser light scattering.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Light-scattering characterization of poly (tetrafluoroethylene). 2. PTFE in perfluorotetracosane: molecular weight distribution and solution properties.
Chu B, et al.
Macromolecules, 22(2), 831-837 (1989)
Nagayama K, et al.
Physica B: Condensed Matter, 208, 419-420 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service