265969
Lithium
rod, diam. 12.7 mm, 99.9% trace metals basis
Synonym(s):
Lithium atom, Lithium element
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Li
CAS Number:
Molecular Weight:
6.94
EC Number:
MDL number:
UNSPSC Code:
12141803
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99.9% trace metals basis
form
rod
reaction suitability
reagent type: reductant
resistivity
9.446 μΩ-cm, 20°C
diam.
12.7 mm
bp
1342 °C (lit.)
mp
180 °C (lit.)
density
0.534 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
[Li]
InChI
1S/Li
InChI key
WHXSMMKQMYFTQS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Our high-purity lithium rod with a diameter of 12.7 mm is a cylindrical piece of metal made of lithium, a highly reactive, silvery-white metal. Lithium is one of the lightest and most electropositive metals, making it highly reactive. Lithium is a good conductor of electricity and heat, has a low melting point (180 °C), and is soft and ductile.
Application
Lithiummetal is widely used to fabricate anode material for rechargeable batteries dueto its low negative electrochemical potential and high theoretical specificcapacity.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B - Water-react 1
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Enikö Zörgö et al.
PLoS genetics, 9(3), e1003388-e1003388 (2013-04-05)
The number of chromosome sets contained within the nucleus of eukaryotic organisms is a fundamental yet evolutionarily poorly characterized genetic variable of life. Here, we mapped the impact of ploidy on the mitotic fitness of baker's yeast and its never
Dongyun Chen et al.
Nanoscale, 5(17), 7890-7896 (2013-07-16)
Two-dimensional nanosheets can leverage on their open architecture to support facile insertion and removal of Li(+) as lithium-ion battery electrode materials. In this study, two two-dimensional nanosheets with complementary functions, namely nitrogen-doped graphene and few-layer WS2, were integrated via a
Sureshbabu Guduguntla et al.
The Journal of organic chemistry, 78(17), 8274-8280 (2013-08-22)
An efficient one-pot synthesis of optically active β-alkyl-substituted alcohols through a tandem copper-catalyzed asymmetric allylic alkylation (AAA) with organolithium reagents and reductive ozonolysis is presented. Furthermore, hydroboration-oxidation following the Cu-catalyzed AAA leads to the corresponding homochiral γ-alkyl-substituted alcohols.
John R Geddes et al.
Lancet (London, England), 381(9878), 1672-1682 (2013-05-15)
We review recent developments in the acute and long-term treatment of bipolar disorder and identify promising future routes to therapeutic innovation. Overall, advances in drug treatment remain quite modest. Antipsychotic drugs are effective in the acute treatment of mania; their
Jian Jiang et al.
Nanoscale, 5(17), 8105-8113 (2013-07-26)
Controlled integration of multiple semiconducting oxides into each single unit of ordered nanotube arrays is highly desired in scientific research for the realization of more attractive applications. We herein report a diffusion-controlled solid-solid route to evolve simplex Co(CO3)0.5(OH)0.11H2O@TiO2 core-shell nanowire
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service