Skip to Content
Merck
All Photos(1)

Documents

264334

Sigma-Aldrich

3,4-Difluorobenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CN
CAS Number:
Molecular Weight:
139.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

52-54 °C (lit.)

SMILES string

Fc1ccc(cc1F)C#N

InChI

1S/C7H3F2N/c8-6-2-1-5(4-10)3-7(6)9/h1-3H

InChI key

BTBFCBQZFMQBNT-UHFFFAOYSA-N

Application

3,4-Difluorobenzonitrile has been used in the preparation of:
  • fluorophenoxy herbicides
  • fluorine substituted benzyl amides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajendra P Tangallapally et al.
Journal of medicinal chemistry, 48(26), 8261-8269 (2005-12-22)
In an ongoing effort to develop new and potent antituberculosis agents, a second-generation series of nitrofuranyl amides was synthesized on the basis of the lead compound 5-nitrofuran-2-carboxylic acid 3,4-dimethoxybenzylamide. The primary design consideration was to improve the solubility and consequently
Synthesis of 3, 4-difluorobenzonitrile and monofluorobenzonitriles by means of halogen-exchange fluorination.
Suzuki H and Kimura Y.
Journal of Fluorine Chemistry, 52(3), 341-351 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service