Skip to Content
Merck
All Photos(1)

Key Documents

263893

Sigma-Aldrich

Phenoxazine

≥99%, purified by sublimation

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NO
CAS Number:
Molecular Weight:
183.21
Beilstein:
143234
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

purified by

sublimation

mp

156-159 °C (lit.)

solubility

benzene: freely soluble(lit.)
chloroform: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
ethanol: freely soluble(lit.)
petroleum ether: very slightly soluble(lit.)

SMILES string

N1c2ccccc2Oc3ccccc13

InChI

1S/C12H9NO/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8,13H

InChI key

TZMSYXZUNZXBOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenoxazine dyes, including several Nile blue analogs, are known to localize selectively in animal tumors.

Application

Phenoxazine is a tricyclic 2′deoxycytidine analog that has been used to improve stacking interactions between heterocycles of oligonucleotide/RNA hybrids and to enhance cellular uptake.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

W M Flanagan et al.
Nature biotechnology, 17(1), 48-52 (1999-01-27)
One of the major barriers to the development of antisense therapeutics has been their poor bioavailability. Numerous oligonucleotide modifications have been synthesized and evaluated for enhanced cellular permeation with limited success. Phenoxazine, a tricyclic 2' deoxycytidine analog, was designed to
C W Lin et al.
Cancer research, 51(4), 1109-1116 (1991-02-15)
The overall goal of our research is to develop effective new photosensitizers for tumor-selective photodynamic therapy. Phenoxazine dyes, including several Nile blue analogues, are known to localize selectively in animal tumors. Structural modifications yielded several series of analogues with substantially
Frédéric Bruyneel et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(33), 8283-8295 (2009-07-23)
Laccases are members of the blue copper oxidases family found in nature. They commonly oxidise a wide range of phenol and aniline derivatives, which in turn are involved in oxidative coupling reactions. Yet, laccases remain rarely described as biocatalysts in
Michael J Rose et al.
Inorganic chemistry, 48(14), 6904-6917 (2009-06-02)
Three ruthenium nitrosyl-dye conjugates, namely, [((OMe)(2)bQb)Ru(NO)(Resf)] (RuNO-Resf), [((OMe)(2)bQb)Ru(NO)(Thnl)] (RuNO-Thnl), and [((OMe)(2)bQb)Ru(NO)(Seln)] (RuNO-Seln) have been synthesized using the tetradentate N4 dicarboxamido ligand H(2)(OMe)(2)bQb. Each nitrosyl of this series is conjugated to a phenoxazine-type heterotricyclic chromophore which has been systematically varied in
Guo-Xiang Li et al.
Journal of biochemical and molecular toxicology, 23(4), 280-286 (2009-08-26)
Phenothiazine (PtzNH) and phenoxazine (PozNH) can protect human erythrocytes against hemolysis induced by 2,2'-azobis(2-amidinopropane hydrochloride) (AAPH), a peroxyl radical supplier. However, an antioxidant may be a pro-oxidant to accelerate the oxidation in the presence of radicals. The aim of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service