Skip to Content
Merck
All Photos(1)

Key Documents

263001

Sigma-Aldrich

Cerium

powder, −40 mesh, 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ce
CAS Number:
Molecular Weight:
140.12
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

form

powder

reaction suitability

reagent type: catalyst
core: cerium

resistivity

73 μΩ-cm, 20°C

particle size

−40 mesh

bp

3443 °C (lit.)

mp

795 °C (lit.)

density

6.67 g/mL at 25 °C (lit.)

SMILES string

[Ce]

InChI

1S/Ce

InChI key

GWXLDORMOJMVQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan M Tomczak et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(3), 904-907 (2013-01-11)
Many inorganic pigments contain heavy metals hazardous to health and environment. Much attention has been devoted to the quest for nontoxic alternatives based on rare-earth elements. However, the computation of colors from first principles is a challenge to electronic structure
David Ecija et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(17), 6678-6681 (2013-04-12)
The tessellation of the Euclidean plane by regular polygons has been contemplated since ancient times and presents intriguing aspects embracing mathematics, art, and crystallography. Significant efforts were devoted to engineer specific 2D interfacial tessellations at the molecular level, but periodic
Joanna M Ogiegło et al.
The journal of physical chemistry. A, 117(12), 2479-2484 (2013-03-09)
The optical properties of gadolinium gallium aluminum garnet, Gd3(Ga,Al)5O12, doped with Ce(3+) are investigated as a function of the Ga/Al ratio, aimed at an improved understanding of the energy flow and luminescence quenching in these materials. A decrease of both
Franziska Schwabe et al.
Chemosphere, 91(4), 512-520 (2013-01-29)
An important aspect in risk assessment of nanoparticles (NPs) is to understand their environmental interactions. We used hydroponic plant cultures to study nanoparticle-plant-root interaction and translocation and exposed wheat and pumpkin to suspensions of uncoated CeO2-NP for 8d (primary particle
Ajay Kumar Yagati et al.
Biosensors & bioelectronics, 47, 385-390 (2013-04-24)
In this study, an enzymatic biosensor for amperometric detection of hydrogen peroxide was developed based on the direct electrochemistry of myoglobin (Mb) on a porous cerium dioxide (CeO2) nanostructured film. The developed film accomplished with large surface area was electrodeposited

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Questions

  1. I am interested in the kind of impurities that may be contained in the Ce metal. Could you please provide me a list of the elements that are contained as impurities?

    1 answer
    1. This product is tested for specific impurities which are listed on the lot-specific Certificate of Analysis. The product has not been tested for any other potential impurities. Please view the sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/COFA/26/263001/263001-BULK_______SHBK2277__.pdf .

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service