Skip to Content
Merck
All Photos(3)

Key Documents

262927

Sigma-Aldrich

Vanadium

turnings, 99.7% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
EC Number:
MDL number:
UNSPSC Code:
12141749
PubChem Substance ID:

vapor pressure

8 mmHg ( 20 °C)

Assay

99.7% trace metals basis

form

turnings

resistivity

24.8-26.0 μΩ-cm, 20°C

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nolan S Horner et al.
Analytica chimica acta, 758, 28-35 (2012-12-19)
A previously developed method based on continuous on-line leaching with artificial gastro-intestinal fluids was used to determine the bio-accessible fraction of As, Cu, Fe, V and Zn in brown and white rice from California by inductively coupled mass spectrometry (ICP-MS).
Shyam Biswas et al.
Physical chemistry chemical physics : PCCP, 15(10), 3552-3561 (2013-02-06)
Two perfluorinated metal hydroxo terephthalates [M(III)(OH)(BDC-F)]·n(guests) (M(III) = V, MIL-47-F-AS or 1-AS; Al, Al-MIL-53-F-AS or 2-AS) (BDC-F = 2-fluoro-1,4-benzenedicarboxylate; AS = as-synthesized) have been synthesized by a hydrothermal method using microwave irradiation (1-AS) or conventional electric heating (2-AS), respectively. The
Biswajit Mukherjee et al.
Toxicology letters, 150(2), 135-143 (2004-04-20)
The biological image of the transition element vanadium ferments a great deal of contradiction-from toxicity to essentiality. Importance of this element as micro-nutrient is yet to be unequivocally accepted by biologists and biomedical scientists. In spite of toxicity, it seems
A V Kudrin
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 14(3), 129-142 (2000-12-29)
In recent years several studies have shown that NF-kappaB might be a very important therapeutic target in the treatment ot various chronic inflammatory, degenerative and tumour diseases. Trace elements play essential roles in the regulation ot cell signaling mechanisms via
Kazuhiro Uehara et al.
Inorganic chemistry, 52(2), 1133-1140 (2013-01-11)
Effects of isolobal heteroatoms in divanadium-substituted γ-Keggin-type polyoxometalates, (TBA)(4)[γ-XV(2)W(10)O(38)(μ-OH)(2)] 1(X) and (TBA)(4)[γ-XV(2)W(10)O(38)(μ-O)] 2(X) (where X = Ge or Si), on (OV)(2)(μ-OH)(2) and (OV)(2)(μ-O) core structures and transformations from 2(X) to 1(X) have been investigated. X-ray crystallography of 1(X) and 2(X)

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service