Skip to Content
Merck
All Photos(1)

Key Documents

260827

Sigma-Aldrich

2-Methylbenzamide

98%

Synonym(s):

o-Toluamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CONH2
CAS Number:
Molecular Weight:
135.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Assay

98%

mp

141-142 °C (lit.)

solubility

alcohol: very soluble(lit.)
benzene: very slightly soluble(lit.)
cold water: insoluble(lit.)
hydrochloric acid: very soluble (conc.)(lit.)
water: very soluble (hot)(lit.)

functional group

amide

SMILES string

Cc1ccccc1C(N)=O

InChI

1S/C8H9NO/c1-6-4-2-3-5-7(6)8(9)10/h2-5H,1H3,(H2,9,10)

InChI key

XXUNIGZDNWWYED-UHFFFAOYSA-N

Application

2-Methylbenzamide (o-Toluamide) has been used in total synthesis of phenolic protoberberine alkaloid, cerasonine[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thanh Nguyen Le et al.
Chemical & pharmaceutical bulletin, 56(7), 1026-1029 (2008-07-02)
First total synthesis of the phenolic protoberberine, cerasonine, was accomplished through a coupling reaction between o-toluamide and benzonitrile. This key step provided the 3-arylisoquinoline which was then successfully converted to 7,8-dihydro-8-oxoprotoberberine through an intramolecular S(N)2 reaction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service