Skip to Content
Merck
All Photos(1)

Key Documents

257214

Sigma-Aldrich

Triisobutylaluminum solution

25 wt. % in toluene

Synonym(s):

TIBA solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]3Al
CAS Number:
Molecular Weight:
198.32
Beilstein:
3587328
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

Quality Level

concentration

25 wt. % in toluene

density

0.848 g/mL at 25 °C

SMILES string

CC(C)C[Al](CC(C)C)CC(C)C

InChI

1S/3C4H9.Al/c3*1-4(2)3;/h3*4H,1H2,2-3H3;

InChI key

MCULRUJILOGHCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:
For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nandita M Weliange et al.
Dalton transactions (Cambridge, England : 2003), 44(34), 15286-15296 (2015-06-05)
The insertion, elimination and isomerisation of octenes with di-n-octylaluminium hydride [HAl(Oct)2], tri-n-octylaluminium [Al(Oct)3] and sec-octylaluminium species have been studied as individual steps in a putative aluminium based contrathermodynamic olefin isomerisation process. While elimination of 1-octene from [Al(Oct)3] is energetically unfavourable
Yunhong Zhang et al.
Carbohydrate polymers, 113, 446-454 (2014-09-27)
Alginate oligosaccharides (AOS), which are marine oligosaccharides, are involved in regulating plant root growth, but the promotion mechanism for AOS remains unclear. Here, AOS (10-80 mg/L) induced the expression of auxin-related gene (OsYUCCA1, OsYUCCA5, OsIAA11 and OsPIN1) in rice (Oryza
Inmaculada Suárez et al.
Journal of chromatography. A, 1407, 193-202 (2015-07-15)
Average molecular weights and polydispersity indexes are some of the most important parameters considered in the polymer characterization. Usually, gel permeation chromatography (GPC) and multi angle light scattering (MALS) are used for this determination, but GPC values are overestimated due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service