Skip to Content
Merck
All Photos(1)

Key Documents

256951

Sigma-Aldrich

Ethylaluminum sesquichloride solution

25 wt. % in toluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2AlCl·Cl2AlC2H5
CAS Number:
Molecular Weight:
212.05
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

concentration

25 wt. % in toluene

SMILES string

CC[Al](Cl)Cl.CC[Al](Cl)CC

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under nitrogen in Sure/Pac™ metal cylinders

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service