Skip to Content
Merck
All Photos(1)

Documents

256919

Sigma-Aldrich

Ethylaluminum dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5AlCl2
CAS Number:
Molecular Weight:
126.95
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

5 mmHg ( 60 °C)

Quality Level

form

solid

bp

115 °C/50 mmHg (lit.)

mp

32 °C (lit.)

density

1.207 g/mL at 25 °C (lit.)

SMILES string

CC[Al](Cl)Cl

InChI

1S/C2H5.Al.2ClH/c1-2;;;/h1H2,2H3;;2*1H/q;+2;;/p-2

InChI key

UAIZDWNSWGTKFZ-UHFFFAOYSA-L

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:
For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Other Notes

Storage below 30°C may cause crystallization. Moving container to a warm location should melt the solid. The warm liquid may be transferred using syringe techniques.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service