Skip to Content
Merck
All Photos(1)

Key Documents

256137

Sigma-Aldrich

2-Butylacrolein

95%

Synonym(s):

2-Methylenehexanal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O
CAS Number:
Molecular Weight:
112.17
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:

Assay

95%

storage temp.

2-8°C

SMILES string

CCCCC(=C)C=O

InChI

1S/C7H12O/c1-3-4-5-7(2)6-8/h6H,2-5H2,1H3

InChI key

IWEWQGKFUYQGSN-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service