Skip to Content
Merck
All Photos(1)

Key Documents

255874

Sigma-Aldrich

Methyl 3-oxovalerate

98%

Synonym(s):

Methyl 3-oxopentanoate, Methyl propionylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COCH2CO2CH3
CAS Number:
Molecular Weight:
130.14
Beilstein:
1753684
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.422 (lit.)

bp

73-74 °C/5 mmHg (lit.)

density

1.037 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC(=O)CC(=O)OC

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service