Skip to Content
Merck
All Photos(1)

Key Documents

255289

Sigma-Aldrich

m-Cymene

99%

Synonym(s):

1-Isopropyl-3-methylbenzene, 3-Isopropyltoluene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$170.00
500 MG
$525.00

$170.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$170.00
500 MG
$525.00

About This Item

Linear Formula:
CH3C6H4CH(CH3)2
CAS Number:
Molecular Weight:
134.22
Beilstein:
1851357
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$170.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor pressure

4 mmHg ( 37.7 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

175 °C (lit.)

mp

−64 °C (lit.)

solubility

alcohol: miscible(lit.)
diethyl ether: miscible(lit.)
water: insoluble(lit.)

density

0.861 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1cccc(C)c1

InChI

1S/C10H14/c1-8(2)10-6-4-5-9(3)7-10/h4-8H,1-3H3

InChI key

XCYJPXQACVEIOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhuiyan Md, et al.
Bangladesh Journal of Pharmacology, 4(2) (2009)
A Ultee et al.
Journal of food protection, 63(5), 620-624 (2000-05-29)
The antimicrobial activity of carvacrol, a compound present in the essential oil fraction of oreganum and thyme, toward the foodborne pathogen Bacillus cereus on rice was studied. Carvacrol showed a dose-related inhibition of growth of the pathogen. Concentrations of 0.15
J W Faller et al.
Chirality, 12(5-6), 325-337 (2000-05-29)
Heterobidentate chelating ligands comprising P,O-donor sets produce chiral metal centers when bound to arene-ruthenium complexes. The cymene complex [CyRu(eta2-chelate-P,O)Cl]+ cations are precursors to 16-electron dicationic strong Lewis acids which have potential use in asymmetric catalysis. Sixteen-electron complexes of this type
[Toxic properties of 1-methyl-3-isopropylbenzene and the substantiation of its MPEL in the air of the work area].
V A Stel'makh
Gigiena truda i professional'nye zabolevaniia, (12)(12), 57-57 (1986-12-01)
Xiu-wei Yang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 31(8), 663-666 (2006-07-13)
To analyze the constituents of the essential oils from Radix Angelicae Pubescentis planted in a GAP base of the Chinese medica materials, and provide the scientific basis for quality control. The essential oils were extracted by water-steam distillation and separated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service