Skip to Content
Merck
All Photos(1)

Key Documents

255254

Sigma-Aldrich

3-Aminobenzophenone

97%

Synonym(s):

3-Benzoylaniline

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$87.50

$87.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$87.50

About This Item

Linear Formula:
H2NC6H4C(O)C6H5
CAS Number:
Molecular Weight:
197.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$87.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

81-84 °C (lit.)

functional group

ketone
phenyl

SMILES string

Nc1cccc(c1)C(=O)c2ccccc2

InChI

1S/C13H11NO/c14-12-8-4-7-11(9-12)13(15)10-5-2-1-3-6-10/h1-9H,14H2

InChI key

FUADXEJBHCKVBN-UHFFFAOYSA-N

General description

3-Aminobenzophenone forms cyclodextrin (α and β) based nanostructures through the supramolecular self assembly[1].

Application

3-Aminobenzophenone has been used in the synthesis of racemic benzophenone ureas, chiral photoaffinity labeling probes[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

N Rajendiran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 127, 52-60 (2014-03-19)
Cyclodextrin (α and β) based nanostructures formed with 2-aminobenzophenone, 3-aminobenzophenone through the supramolecular self assembly are studied by absorption, fluorescence, time-resolved fluorescence, SEM, TEM, FT-IR, DSC, PXRD and (1)H NMR. The unequal layer by layer nanosheets and nanoribbons are formed
Elizabeth M Hadac et al.
Journal of medicinal chemistry, 49(3), 850-863 (2006-02-03)
An understanding of the molecular basis of drug action provides opportunities for refinement of drug properties and for development of more potent and selective molecules that act at the same biological target. In this work, we have identified the active

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service