Skip to Content
Merck
All Photos(2)

Key Documents

254568

Sigma-Aldrich

O-Allylhydroxylamine hydrochloride hydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2ONH2·HCl·xH2O
CAS Number:
Molecular Weight:
109.55 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

mp

175 °C (dec.) (lit.)

solubility

methanol: soluble 25 mg/mL, clear, colorless

SMILES string

Cl[H].[H]O[H].NOCC=C

InChI

1S/C3H7NO.ClH.H2O/c1-2-3-5-4;;/h2H,1,3-4H2;1H;1H2

InChI key

VYCHJEXJHFTAQI-UHFFFAOYSA-N

Application

O-Allylhydroxylamine hydrochloride hydrate has been used in the preparation of acetophenone O-allyloxime[1] and O-allyI-N-(2-trimethylsilylethyloxycarbonyl)-hydroxylamine[2].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of (R)-{η 6-[O-methyl-N-(α-methylbenzyl) hydroxyamino] benzene} chromium tricarbonyl via nucleophilic aromatic substitution of (η 6-fluorobenzene) chromium tricarbonyl.
da Costa MRG, et al.
Journal of the Chemical Society. Perkin Transactions 1, 21, 2850-2855 (2001)
J H van Maarseveen et al.
Bioorganic & medicinal chemistry, 5(5), 955-970 (1997-05-01)
In a search for the minimum pharmacophore of the naturally occurring tetracyclic eudistomins, five structural analogues (4-8) were evaluated for their in vitro antiviral and tumor cell antiproliferative activities. For the synthesis of these derivatives both intra- and intermolecular Pictet-Spengler

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service