Skip to Content
Merck
All Photos(1)

Key Documents

254509

Sigma-Aldrich

Trimethyl orthobutyrate

97%

Synonym(s):

1,1,1-Trimethoxybutane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$221.00

$221.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$221.00

About This Item

Linear Formula:
CH3CH2CH2C(OCH3)3
CAS Number:
Molecular Weight:
148.20
Beilstein:
1737319
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$221.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.404 (lit.)

bp

145-147 °C (lit.)

density

0.926 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCCC(OC)(OC)OC

InChI

1S/C7H16O3/c1-5-6-7(8-2,9-3)10-4/h5-6H2,1-4H3

InChI key

JAFMOTJMRSZOJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics and mechanism of gas-phase elimination of trimethyl orthobutyrate has been examined over the temperature range of 310-369°C and pressure range of 50-130Torr[1].

Application

Trimethyl orthobutyrate has been used in the preparation of:
  • 5-acetamido-9-O-butyroyl-3,5-dideoxy-D-glycero-2-nonulo pyranosonic acid[2]
  • 9-O-butyroyl-3,5-dideoxy-5-glycoloylamido-D-glycero-2-nonulo pyranosonic acid[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ogura et al.
Carbohydrate research, 167, 77-86 (1987-09-15)
Various 9-O-acyl derivatives of N-acetyl- and N-glycoloyl-neuraminic acid, and O-(5-acetamido-3,5-dideoxy-D-glycero-alpha- and beta-D-galacto-2-nonulopyranosylonic acid)-(2----6)-O-beta-D-galactopyranosyl-(1----4)-D-glucopyranose were regioselectively synthesized by use of ortho esters. In addition, 5-acetamido-4-O-acetyl-D-glycero-D-galacto-2-nonulopyranosonic acid was prepared starting from the benzyl and methyl esters of N-acetylneuraminic acid.
Edgar Márquez et al.
The journal of physical chemistry. A, 112(47), 12140-12142 (2008-11-01)
The gas-phase elimination kinetics of the title compounds have been examined over the temperature range of 310-369 degrees C and pressure range of 50-130 Torr. The reactions, in seasoned vessels, are homogeneous, unimolecular, and follow a first-order rate law. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service