Skip to Content
Merck
All Photos(1)

Key Documents

252131

Sigma-Aldrich

1,5-Diiodopentane

97%, contains copper as stabilizer

Synonym(s):

α,ψ-Diiodopentane, (±)-Hexane-1,2-diol, Pentamethylene diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I(CH2)5I
CAS Number:
Molecular Weight:
323.94
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.6002 (lit.)

bp

101-102 °C/3 mmHg (lit.)

density

2.177 g/mL at 25 °C (lit.)

functional group

iodo

SMILES string

ICCCCCI

InChI

1S/C5H10I2/c6-4-2-1-3-5-7/h1-5H2

InChI key

IAEOYUUPFYJXHN-UHFFFAOYSA-N

General description

Electrochemical reduction of 1,5-diiodopentane at carbon electrodes in DMF containing tetramethylammonium perchlorate has been studied by cyclic voltammetry and controlled-potential electrolysis[1].

Application

1,5-Diiodopentane has been used:
  • as crosslinking reagent, to investigate the cross-linking layer-by-layer films of bioreducible poly(2-dimethylaminoethyl methacrylate) and DNA[2]
  • in preparation of (iodoalkylaminocarbene)tungsten complexes[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of a-lithio (aminocarbene) tungsten anions with diiodoalkanes: Synthesis of (?-bis (aminocarbene)) ditungsten complexes.
Macomber DW and Madhukar P.
Journal of Organometallic Chemistry, 433(3), 279-285 (1992)
Jenifer Blacklock et al.
The journal of physical chemistry. B, 114(16), 5283-5291 (2010-04-08)
The effect of cross-linking layer-by-layer (LbL) films consisting of bioreducible poly(2-dimethylaminoethyl methacrylate) (rPDMAEMA) and DNA is examined with regard to rigidity, biodegradability, cell adhesion, and transfection activity using 1,5-diiodopentane (DIP) cross-linker. DIP chemically reacts with the tertiary amines of rPDMAEMA
Electrochemical Reduction of 1, 5-Dihalopentanes at Carbon Cathodes in Dimethylformamide.
Pritts WA and Peters DG.
Journal of the Electrochemical Society, 141(12), 3318-3324 (1994)
Ian C Watson et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(41), 9678-9690 (2019-05-16)
New N-heterocyclic olefins (NHOs) are described with functionalization on the ligand heterocyclic backbone and terminal alkylidene positions. Various PdII -NHO complexes have been formed and their use as pre-catalysts in Buchwald-Hartwig aminations was explored. The most active system for catalytic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service