Skip to Content
Merck
All Photos(2)

Documents

251429

Sigma-Aldrich

Rubidium fluoride

99.8% trace metals basis

Synonym(s):

Rubidium monofluoride, Rubidium(I) fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbF
CAS Number:
Molecular Weight:
104.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.8% trace metals basis

form

solid

impurities

≤1% HF

bp

1410 °C/1 atm (lit.)

mp

795 °C (lit.)

solubility

ethanol: insoluble(lit.)

density

3.557 g/mL at 25 °C (lit.)

SMILES string

[F-].[Rb+]

InChI

1S/FH.Rb/h1H;/q;+1/p-1

InChI key

AHLATJUETSFVIM-UHFFFAOYSA-M

Related Categories

Application

  • Rubidium Fluoride Post-Deposition Treatment: Impact on the Chemical Structure of the Cu(In,Ga)Se2 Thin-Film Surface and the CdS/Cu(In,Ga)Se2 Interface: This study explores the effects of RbF post-deposition treatment on Cu(In,Ga)Se2 thin films and their interfaces with CdS, highlighting changes in chemical composition and electronic properties (Kreikemeyer-Lorenzo et al., 2018).
  • Rubidium Fluoride Additive for High-Efficiency and Low-Hysteresis All-Inorganic CsPbI3 Perovskite Solar Cells: This research introduces RbF as an additive in perovskite solar cells, significantly enhancing efficiency and reducing hysteresis (Zhang et al., 2023).

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service