Skip to Content
Merck
All Photos(1)

Documents

250554

Sigma-Aldrich

cis-2-Methoxycinnamic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
178.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

94-96 °C (lit.)

SMILES string

COc1ccccc1\C=C/C(O)=O

InChI

1S/C10H10O3/c1-13-9-5-3-2-4-8(9)6-7-10(11)12/h2-7H,1H3,(H,11,12)/b7-6-

InChI key

FEGVSPGUHMGGBO-SREVYHEPSA-N

Related Categories

General description

cis-2-Methoxycinnamic acid is the major component of essential oil extracted from Cinnamomum cassia.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-Tze Chou et al.
International journal of molecular sciences, 14(9), 19186-19201 (2013-09-21)
Essential oils extracted from aromatic plants exhibit important biological activities and have become increasingly important for the development of aromatherapy for complementary and alternative medicine. The essential oil extracted from Cinnamomum cassia Presl (CC-EO) has various functional properties; however, little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service