Skip to Content
Merck
All Photos(1)

Key Documents

250406

Sigma-Aldrich

2-(Methylthio)-2-thiazoline

97%

Synonym(s):

2-(Methylmercapto)-2-thiazoline

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$211.00

$211.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$211.00

About This Item

Empirical Formula (Hill Notation):
C4H7NS2
CAS Number:
Molecular Weight:
133.24
Beilstein:
106353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$211.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.592 (lit.)

bp

216-217 °C (lit.)

density

1.226 g/mL at 25 °C (lit.)

SMILES string

CSC1=NCCS1

InChI

1S/C4H7NS2/c1-6-4-5-2-3-7-4/h2-3H2,1H3

InChI key

QFGRBBWYHIYNIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(Methylthio)-2-thiazoline has been used in the preparation of:
  • novel tri- and tetracyclic hetero systems, via reaction with heteroaromatic 2-aminoesters[1]
  • 2-(2-chlorobenzylamino)-2-thiazoline[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reagents for new heteroannelation reactions III. 2-(Methylthio)-2-thiazoline.
Sauter F, et al.
Monatshefte fur Chemie / Chemical Monthly, 128(5), 503-508 (1997)
Synthesis and octopaminergic agonist activity of 2-(substituted benzylamino)-2-thiazolines.
Hirashima A, et al.
Bioscience, Biotechnology, and Biochemistry, 56, 1062-1062 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service