Skip to Content
Merck
All Photos(3)

Key Documents

250171

Sigma-Aldrich

Sodium hexafluorosilicate

Synonym(s):

Disodium hexafluorosilicon(2-), Sodium fluosilicate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SiF6
CAS Number:
Molecular Weight:
188.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

form

powder

Quality Level

density

2.68 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].F[Si--](F)(F)(F)(F)F

InChI

1S/F6Si.2Na/c1-7(2,3,4,5)6;;/q-2;2*+1

InChI key

TWGUZEUZLCYTCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L C Chow et al.
Caries research, 36(2), 136-141 (2002-05-31)
A previous study showed that a sodium hexafluorosilicate-calcium chloride-based two-solution fluoride (F) rinse containing 6 mmol/l of F was more effective than a 12 mmol/l F sodium fluoride rinse in depositing F on tooth surfaces and increasing oral F levels.
P S Lakshminarayanan et al.
Inorganic chemistry, 45(11), 4372-4380 (2006-05-23)
Synthesis and crystal structure of a tren-based amide, L(1), N,N',N''-tris[(2-amino-ethyl)-3-nitro-benzamide] is reported. The crystallographic results show intramolecular hydrogen bonding and aromatic pi...pi stacking among tripodal arms which prevent opening of the receptor cavity. Intermolecular hydrogen bonding in L1 generates the
Je Seon Song et al.
Anticancer research, 25(1A), 391-395 (2005-04-09)
Fluorine compounds are widely used for the prevention of caries, and recently sodium fluorosilicate has been used in water fluorination. The cytotoxic effects of sodium fluorosilicate in several osteosarcoma and oral cancer cells were evaluated in this study by measurement
Shingo Shibata et al.
American journal of dentistry, 24(3), 148-152 (2011-08-31)
To evaluate the degree of penetration of an ammonium hexafluorosilicate [SiF: (NH4)2SiF6] solution containing various antibacterial agents into dentin and the depth of dentin tubule occlusion by the precipitate. Various antibacterial SiF solutions were prepared with the addition to chlorhexidine
Ali Ouasri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(4), 851-857 (2003-03-01)
The X-ray powder diffraction pattern of [(CH(3))(3)NH](2)SiF(6) was obtained and indexed on the basis of a centred cubic unit cell with the P4(1)32 as the likely space group. The Infrared and Raman spectra of this compound have been recorded at

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service