Skip to Content
Merck
All Photos(1)

Documents

24898

Sigma-Aldrich

Chloride on polymer support

extent of labeling: ~4 mmol/g chloride loading

Synonym(s):

Amberlite IRA 900 Cl--form

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

~4 mmol/g chloride loading

impurities

~10% water

particle size

20-50 mesh

Other Notes

Reagent for substitution reactions

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Cainelli et al.
Synthesis, 472-472 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service