Skip to Content
Merck
All Photos(3)

Key Documents

247375

Sigma-Aldrich

Boron tribromide dimethyl sulfide complex

97%

Synonym(s):

Dimethyl sulfide-tribromoborane, Tribromoborane-methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BBr3 · S(CH3)2
CAS Number:
Molecular Weight:
312.66
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

106-108 °C (lit.)

functional group

thioether

SMILES string

C[S+](C)[B-](Br)(Br)Br

InChI

1S/C2H6BBr3S/c1-7(2)3(4,5)6/h1-2H3

InChI key

NCVLHAUANAMSTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Boron tribromide dimethyl sulfide complex (BBr3S(CH3)2) can be used as a reagent:
  • To deprotect the methylenedioxy acetal bonding of poly vinyl-1, 3-benzodioxole.
  • To prepare benzyl S,S′-diphenyl acetal by reacting with carboxylic acid and thexylphenylthioborane.
  • To prepare Baylis-Hillman adducts by the reaction of aldehydes with 3-buten-2-one.

Boron tribromide in convenient solid form. Used as a brominating agent.
Reactant for preparation of:
  • Precursor of HIV protease inhibitor KNI 764 using cation-exchange resin mediated Mannich reaction

Reagent for:
  • Baylis-Hillman reactions

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protection and Polymerization of Functional Monomers. 24. Anionic Living Polymerizations of 5-Vinyl-and 4-Vinyl-1, 3-benzodioxoles
Ishizone T, et al.
Macromolecules, 28(11), 3787-3793 (1995)
Inorganic Chemistry, 29, 4162-4162 (1990)
Direct conversion of carboxylic acids and carboxylic esters into S, S?-diphenyl acetals and phenyl sulfides with thexylphenylthioborane
Kim S and Kim SS
Tetrahedron Letters, 28(17), 1913-1916 (1987)
Dimethyl sulfide-boron trihalide-mediated reactions of α, β, unsaturated ketones with aldehydes: one-pot synthesis of Baylis-Hillman adducts and ?-halomethyl enones
Iwamura T, et al.
Tetrahedron, 57(40), 8455-8462 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service