Skip to Content
Merck
All Photos(1)

Key Documents

246875

Sigma-Aldrich

4-Cyclopentylphenol

technical grade, 95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$84.20

$84.20


Estimated to ship onApril 30, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$84.20

About This Item

Linear Formula:
C5H9C6H4OH
CAS Number:
Molecular Weight:
162.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$84.20


Estimated to ship onApril 30, 2025


Request a Bulk Order

grade

technical grade

Quality Level

Assay

95%

bp

155 °C/12 mmHg (lit.)

mp

64-66 °C (lit.)

SMILES string

Oc1ccc(cc1)C2CCCC2

InChI

1S/C11H14O/c12-11-7-5-10(6-8-11)9-3-1-2-4-9/h5-9,12H,1-4H2

InChI key

SNBKPVVDUBFDEJ-UHFFFAOYSA-N

General description

Resolution of 2-, 3- and 4-cyclopentylphenol (positional isomers of cyclopentylphenol) by thin-layer chromatography has been reported[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Separation of 2-, 3-and 4-Cyclopentylphenol by Thin-Layer Chromatography.
Atkins TM, et al.
Journal of Liquid Chromatography, 6(7), 1171-1174 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service