Skip to Content
Merck
All Photos(1)

Key Documents

245542

Sigma-Aldrich

4,4-Dimethyl-2-cyclohexen-1-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H6(=O)
CAS Number:
Molecular Weight:
124.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.473 (lit.)

bp

72-73.5 °C/20 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC1(C)CCC(=O)C=C1

InChI

1S/C8H12O/c1-8(2)5-3-7(9)4-6-8/h3,5H,4,6H2,1-2H3

InChI key

HAUNPYVLVAIUOO-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guifang Zhang et al.
Investigative ophthalmology & visual science, 48(11), 5243-5249 (2007-10-27)
To describe a method of drug delivery to the retina via aerosolized nanoparticles in the gas phase during the gas-exchange stage of vitrectomy in a porcine model. An ultrasonically atomized and dried sodium fluorescein aerosol was produced with a concentration
Studies on the reactions of α, β-enones with allyl indium reagent; effects of TMSCl as promoter on regioselectivity.
Lee PH, et al.
Tetrahedron Letters, 42(1), 37-39 (2001)
Borja Oliver-Tomas et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(52), 12900-12908 (2017-07-06)
For the reaction mechanism of the ketonic decarboxylation of two carboxylic acids, a β-keto acid is favored as key intermediate in many experimental and theoretical studies. Hydrogen atoms in the α-position are an indispensable requirement for the substrates to react

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service