Skip to Content
Merck
All Photos(2)

Documents

242802

Sigma-Aldrich

Sulfoacetic acid

technical grade

Synonym(s):

2-Sulfoacetic acid, Carboxymethylsulfonic acid, Sulfoethanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO3SCH2CO2H
CAS Number:
Molecular Weight:
140.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

mp

81-88 °C (lit.)

SMILES string

OC(=O)CS(O)(=O)=O

InChI

1S/C2H4O5S/c3-2(4)1-8(5,6)7/h1H2,(H,3,4)(H,5,6,7)

InChI key

AGGIJOLULBJGTQ-UHFFFAOYSA-N

Application

Sulfoacetic acid was used in the synthesis of ethyl ester by esterification in refluxing ethanol.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H T Lee et al.
Bioorganic & medicinal chemistry letters, 8(3), 289-294 (1999-01-01)
Sulfoacetic acid, phosphoramidate, and phosphoramide analogs of the ACAT inhibitors, CI-999 and CI-1011 were synthesized. The structure-activity relationships of these compounds as ACAT inhibitors are described.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service