236942
Dichloromethane-d2
99.5 atom % D
Synonym(s):
Dideuteromethylenechloride, Methylene chloride-d2
About This Item
vapor pressure
24.77 psi ( 55 °C)
7.02 psi ( 20 °C)
Quality Level
isotopic purity
99.5 atom % D
packaging
pk of 10 × 1 mL
refractive index
n20/D 1.422 (lit.)
bp
40 °C (lit.)
density
1.362 g/mL at 25 °C (lit.)
mass shift
M+2
SMILES string
[2H]C([2H])(Cl)Cl
InChI
1S/CH2Cl2/c2-1-3/h1H2/i1D2
InChI key
YMWUJEATGCHHMB-DICFDUPASA-N
Looking for similar products? Visit Product Comparison Guide
accessory
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service