Skip to Content
Merck
All Photos(1)

Documents

236357

Sigma-Aldrich

3-Hydroxypropionitrile

ReagentPlus®, 99%

Synonym(s):

2-Cyanoethanol, Ethylene cyanohydrin, Hydracrylonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2CN
CAS Number:
Molecular Weight:
71.08
Beilstein:
635773
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

922 °F

expl. lim.

12.1 %

refractive index

n20/D 1.425 (lit.)

bp

228 °C (lit.)

mp

−46 °C (lit.)

solubility

diethyl ether: slightly soluble 2.3 % (w/w) at 15 °C(lit.)
acetone: miscible(lit.)
benzene: insoluble(lit.)
carbon disulfide: insoluble(lit.)
carbon tetrachloride: insoluble(lit.)
ethanol: miscible(lit.)
petroleum ether: insoluble(lit.)
water: miscible(lit.)

density

1.04 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCCC#N

InChI

1S/C3H5NO/c4-2-1-3-5/h5H,1,3H2

InChI key

WSGYTJNNHPZFKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3-Hydroxypropionitrile has been used:
  • as polar solvent to investigate the orientation of the phospholipid at the surface of the polar solvents by neutral impact collision ion scattering spectroscopy
  • in solid state composite electrolyte for dye-sensitized solar cells
  • as a reagent for carboxyl protection in peptide synthesis

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongxia Wang et al.
The journal of physical chemistry. B, 110(12), 5970-5974 (2006-03-24)
It was observed that the ionic conductivity of the solid-state electrolyte LiI/3-hydroxypropionitrile (HPN) = 1:4 (molar ratio) decreased dramatically with increasing iodine (I(2)) concentration, which differs from the conduction behavior of the Grotthuss transport mechanism observed in liquid or gel
3-hydroxypropionitrile: A new reagent for carboxyl protection in peptide synthesis.
Misra PK, et al.
Tetrahedron Letters, 30(27), 3569-3572 (1989)
Hartwig Pohl et al.
Langmuir : the ACS journal of surfaces and colloids, 26(4), 2473-2476 (2009-11-20)
Phospholipids are a main component of cell membranes. Therefore, the experimental investigation of the self-organization of phospholipids is of great interest. Here we present results concerning the orientation of the phospholipid 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC) at the surface of the polar solvents
Kid Törnquist et al.
European journal of pharmacology, 453(1), 1-11 (2002-10-24)
Sphingolipid derivatives cause diverse effects towards the regulation of intracellular free Ca(2+) concentrations ([Ca(2+)](i)) in a multitude of nonexcitable cells. In the present investigation, the effect of C-8 ceramide-1-(2-cyanoethyl) phosphate (C1CP) on store-operated Ca(2+) (SOC) entry was investigated. C1CP evoked
M Duverger-Van Bogaert et al.
Toxicology letters, 7(4-5), 311-319 (1981-02-01)
The mutagenicity of acrylonitrile (ACN) was tested with Salmonella typhimurium TA1530 after a preincubation period of the chemical with a rat liver post-mitochondrial fraction in liquid medium. Several pretreatments were applied to the animals before the preparation of the liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service