Skip to Content
Merck
All Photos(3)

Documents

235849

Sigma-Aldrich

Aluminum tert-butoxide

technical grade

Synonym(s):

Aluminum-tri-tert-butoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al[OC(CH3)3]3
CAS Number:
Molecular Weight:
246.32
Beilstein:
4148229
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

form

powder

composition

Al, 10.5-13.0% EDTA titration

reaction suitability

core: aluminum

SMILES string

CC(C)(C)O[Al](OC(C)(C)C)OC(C)(C)C

InChI

1S/3C4H9O.Al/c3*1-4(2,3)5;/h3*1-3H3;/q3*-1;+3

InChI key

MDDPTCUZZASZIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service