Skip to Content
Merck
All Photos(3)

Key Documents

234249

Sigma-Aldrich

Guanidine nitrate

98%

Synonym(s):

Guanidinium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HNO3
CAS Number:
Molecular Weight:
122.08
Beilstein:
3596600
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
98%
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

mp

213-215 °C (lit.)

solubility

water: soluble 50 mg/mL, clear to very slightly hazy, colorless

functional group

amine
nitrate

SMILES string

NC(N)=N.O[N+]([O-])=O

InChI

1S/CH5N3.HNO3/c2*2-1(3)4/h(H5,2,3,4);(H,2,3,4)

InChI key

CNUNWZZSUJPAHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Guanidine nitrate (GN) is an explosive used by the military and commercial sectors[1]. Decomposition of guanidine nitrate has been investigated[1].

Application

Guanidine nitrate has been used:
  • in ammonium nitrate explosive formulations and in various propellant applications including triple-base gun propellants[1]
  • to examine the vapor signatures of the nitrate salts of guanidine by isothermal thermogravimetric method[2]

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Decompositions of urea and guanidine nitrates.
Oxley JC, et al.
Journal of Energetic Materials, 27(1), 17-39 (2008)
Yunfei Zhou et al.
Environmental science and pollution research international, 26(10), 9861-9875 (2019-02-09)
Montmorillonite grafted polyacrylic acid composite (GNM) was prepared by using ultraviolet radiation grafting method in this work. The synthesized materials were characterized by XRF, SEM, FTIR, XRD, TG, and XPS. The experimental equilibrium data indicates that the adsorbent is suitable
Yeon-Mi Lim et al.
Toxics, 8(3) (2020-09-26)
The toxicity profiles of the widely used guanidine-based chemicals have not been fully elucidated. Herein, we evaluated the in vitro and in vivo toxicity of eight guanidine-based chemicals, focusing on inhalation toxicity. Among the eight chemicals, dodecylguanidine hydrochloride (DGH) was
Determination of urea nitrate and guanidine nitrate vapor pressures by isothermal thermogravimetry.
Oxley J, et al.
Propellants, Explosives, Pyrotechnics, 35(3), 278-283 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service