Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

233919

Sigma-Aldrich

Lithium D-lactate

98%

Synonym(s):

(R)-2-Hydroxypropionic acid lithium salt, D-Lactic acid lithium salt

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
CH3CH(OH)CO2Li
CAS Number:
Molecular Weight:
96.01
Beilstein:
4823618
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

optical activity

[α]20/D +14.3°, c = 1 in H2O

SMILES string

[Li+].C[C@@H](O)C([O-])=O

InChI

1S/C3H6O3.Li/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m1./s1

InChI key

GKQWYZBANWAFMQ-HSHFZTNMSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service