Skip to Content
Merck
All Photos(1)

Documents

232890

Sigma-Aldrich

2′,6′-Difluoroacetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3COCH3
CAS Number:
Molecular Weight:
156.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.48 (lit.)

bp

76-79 °C/15 mmHg (lit.)

density

1.197 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1c(F)cccc1F

InChI

1S/C8H6F2O/c1-5(11)8-6(9)3-2-4-7(8)10/h2-4H,1H3

InChI key

VGIIILXIQLXVLC-UHFFFAOYSA-N

General description

2′,6′-Difluoroacetophenone undergoes ruthenium-catalyzed phenylation with phenylboronate via carbon-fluorine bond cleavage to yield 2′,6′-diphenylacetophenone.

Application

2′,6′-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium-catalyzed arylation of fluorinated aromatic ketones via< i> ortho</i>-selective carbon-fluorine bond cleavage.
Kawamoto K, et al.
Tetrahedron Letters, 52(44), 5888-5890 (2011)
Synthesis of 2-aminoquinazolines from ortho-fluoroketones.
Hynes JB, et al.
Journal of Heterocyclic Chemistry, 32(4), 1185-1187 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service