Skip to Content
Merck
All Photos(3)

Key Documents

230162

Sigma-Aldrich

Sulforhodamine B

Dye content 75 %

Synonym(s):

Acid Red 52, Sulforhodamine B monosodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$101.00
5 G
$316.00

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$101.00
5 G
$316.00

About This Item

Empirical Formula (Hill Notation):
C27H29N2NaO7S2
CAS Number:
Molecular Weight:
580.65
Colour Index Number:
45100
Beilstein:
3886008
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥73% (HPLC)

Quality Level

form

powder

composition

Dye content, 75%

technique(s)

microbe id | staining: suitable

color

brown to very dark brown
green to very dark green

solubility

methanol: 1 mg/mL

λmax

554 nm

ε (extinction coefficient)

≥85000 at 554-560 nm in 0.1 M NaOH in methanol at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CCN(CC)c1ccc2c(OC3=CC(\C=CC3=C2c4ccc(cc4S([O-])(=O)=O)S([O-])(=O)=O)=[N+](\CC)CC)c1

InChI

1S/C27H30N2O7S2.Na/c1-5-28(6-2)18-9-12-21-24(15-18)36-25-16-19(29(7-3)8-4)10-13-22(25)27(21)23-14-11-20(37(30,31)32)17-26(23)38(33,34)35;/h9-17H,5-8H2,1-4H3,(H-,30,31,32,33,34,35);/q;+1/p-1

InChI key

SXQCTESRRZBPHJ-UHFFFAOYSA-M

General description

Sulforhodamine B (SRB) is a bright-pink aminoxanthene dye. The sulforhodamine B (SRB) assay is extensively used for in vitro cytotoxicity screening. SRB binds to the protein constituents of the cells in a stoichiometric manner. Thus, the amount of dye extracted from stained cells is directly proportional to the cell mass. SRB can also be used for cell density determination.[1]

Application

Sulforhodamine B has been used as a dye in the MCF-7 (human breast adenocarcinoma cell line) cell proliferation assay.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endocrine disrupting effects in vitro of conazole antifungals used as pesticides and pharmaceuticals
Kj?rstad MB
Reproductive Toxicology, 30, 573-582 (2010)
Sulforhodamine B colorimetric assay for cytotoxicity screening
Vichai V
Nature Protocols, 1, 1112?1116-1112?1116 (2006)
Christian A Ruge et al.
Colloids and surfaces. B, Biointerfaces, 139, 219-227 (2016-01-01)
The conversion of colloidal drug carriers/polymeric nanoparticles into dry microparticulate powders (e.g., by spray-drying) is a prominent approach to overcome the aerodynamic limitations of these formulations for delivery via inhalation. However, to what extent such nano-embedded microparticles disintegrate into individual/intact
Samantha N Andrews et al.
Pharmaceutical research, 30(4), 1099-1109 (2012-12-01)
Most methods to increase transdermal drug delivery focus on increasing stratum corneum permeability, without addressing the need to increase permeability of viable epidermis. Here, we assess the hypothesis that viable epidermis offers a significant permeability barrier that becomes rate limiting
Anna Jaśkiewicz et al.
Journal of clinical medicine, 8(5) (2019-05-19)
The present study is centered on molecular mechanisms of the cytoprotective effect of geranylgeraniol (GGOH) in skeletal muscle harmed by statin-associated myopathy (SAM). GGOH via autophagy induction was purportedly assumed to prevent skeletal muscle viability impaired by statins, atorvastatin (ATR)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service