Skip to Content
Merck
All Photos(1)

Documents

228192

Sigma-Aldrich

Dibutyltin bis(2-ethylhexanoate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CO2]2Sn[(CH2)3CH3]2
CAS Number:
Molecular Weight:
519.35
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

57-59 °C (lit.)

SMILES string

CCCCC(CC)C(=O)O[Sn](CCCC)(CCCC)OC(=O)C(CC)CCCC

InChI

1S/2C8H16O2.2C4H9.Sn/c2*1-3-5-6-7(4-2)8(9)10;2*1-3-4-2;/h2*7H,3-6H2,1-2H3,(H,9,10);2*1,3-4H2,2H3;/q;;;;+2/p-2

InChI key

GPDWNEFHGANACG-UHFFFAOYSA-L

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Repr. 1B - Skin Corr. 1C - Skin Sens. 1 - STOT RE 1

Target Organs

Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

244.4 °F

Flash Point(C)

118 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service