Skip to Content
Merck
All Photos(1)

Key Documents

22669

Sigma-Aldrich

Fmoc-Phe(4-Boc2-guanidino)-OH

technical, ≥90% (HPLC)

Synonym(s):

Fmoc-4-(N,N′-di-Boc-guanidino)-L-phenylalanine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$347.00

$347.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$347.00

About This Item

Empirical Formula (Hill Notation):
C35H40N4O8
CAS Number:
Molecular Weight:
644.71
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$347.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical

Quality Level

Assay

≥90% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N\C(NC(=O)OC(C)(C)C)=N/c1ccc(C[C@H](NC(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O)cc1

InChI

1S/C35H40N4O8/c1-34(2,3)46-32(43)38-30(39-33(44)47-35(4,5)6)36-22-17-15-21(16-18-22)19-28(29(40)41)37-31(42)45-20-27-25-13-9-7-11-23(25)24-12-8-10-14-26(24)27/h7-18,27-28H,19-20H2,1-6H3,(H,37,42)(H,40,41)(H2,36,38,39,43,44)/t28-/m0/s1

InChI key

CIEGZABLYKOZLG-NDEPHWFRSA-N

Other Notes

Pharmacologically interesting protected analog of arginine.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Melo et al.
Biochemistry, 40(17), 5226-5232 (2001-04-25)
We explored the unique substrate specificity of the primary S(1) subsite of human urinary kallikrein (hK1), which accepts both Phe and Arg, using internally quenched fluorescent peptides Abz-F-X-S-R-Q-EDDnp and Abz-G-F-S-P-F-X-S-S-R-P-Q-EDDnp [Abz is o-aminobenzoic acid; EDDnp is N-(2,4-dinitrophenyl)ethylenediamine], which were based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service