Skip to Content
Merck
All Photos(1)

Key Documents

226130

Sigma-Aldrich

3-Bromo-1,2-propanediol

97%

Synonym(s):

α-Glycerol bromohydrin

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$139.00
25 G
$479.00

$139.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$139.00
25 G
$479.00

About This Item

Linear Formula:
CH2BrCHOHCH2OH
CAS Number:
Molecular Weight:
154.99
Beilstein:
1719124
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$139.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.518 (lit.)

bp

72-75 °C/0.2 mmHg (lit.)

density

1.771 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC(O)CBr

InChI

1S/C3H7BrO2/c4-1-3(6)2-5/h3,5-6H,1-2H2

InChI key

SIBFQOUHOCRXDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Protecting reagent for carbonyl functions.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Jones et al.
Journal of applied toxicology : JAT, 16(1), 57-63 (1996-01-01)
(R,S)- and (R)-3-Bromopropan-1,2-diol (alpha-bromohydrin) produced diuresis and glucosuria when administered to male rats but had no effect on the metabolic activity of rat kidney tubules in vitro. The oxidation products (R,S)-3-bromolactate and 3-bromopyruvate produced brief phases of diuresis but not
A R Jones et al.
Journal of reproduction and fertility, 108(1), 95-100 (1996-09-01)
Boar spermatozoa incubated with glycerol 3-phosphate as substrate produced CO2 and an accumulation of dihydroxyacetone phosphate and fructose-1,6-bisphosphate. The rate of oxidation of glycerol 3-phosphate was decreased, as was the production of CO2 and the two glycolytic intermediates, in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service