Skip to Content
Merck
All Photos(1)

Key Documents

225452

Sigma-Aldrich

Bis(triphenylphosphine)palladium(II) diacetate

98%

Synonym(s):

Bis(acetato)bis(triphenylphosphine)palladium(II), Palladium(II)bis(triphenylphosphine) diacetate, Pd(OAc)2(PPh3)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]2Pd(CH3COO)2
CAS Number:
Molecular Weight:
749.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

136 °C (dec.) (lit.)

SMILES string

CC(=O)O[Pd]OC(C)=O.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.2C2H4O2.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;2*1-2(3)4;/h2*1-15H;2*1H3,(H,3,4);/q;;;;+2/p-2

InChI key

UVBXZOISXNZBLY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Bis(triphenylphosphine)palladium(II) diacetate can be used as a catalyst for C-C bond formation via Sonogashira coupling, Negishi coupling, Heck coupling, and Suzuki coupling reaction.[1][2][3]
It can also be used as a catalyst to synthesize:
  • Pyrido[1,2-a]benzimidazole derivatives by electro-oxidative intramolecular C-H/N-H annulation reaction.[3]
  • α, β-Unsaturated carboxylic acids by hydroxycarbonylation of vinyl triflates.[4]
  • Secondary imides by C-H functionalization of aldehydes with different N-substituted N-heteroarene-2-carboxamides.[5]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Easy-separable magnetic nanoparticle-supported Pd catalysts: Kinetics, stability and catalyst re-use
Laska U, et al.
J. Catal., 268(2), 318-328 (2009)
Palladium-Catalyzed Electro-oxidative C-H Amination toward the Synthesis of Pyrido [1, 2-a] benzimidazoles with Hydrogen Evolution
Duan Z, et al.
ACS Catalysis, 10(6), 3828-3831 (2020)
Yus, M.; Gomis, J.
Tetrahedron Letters, 42, 5721-5721 (2001)
Cross coupling reactions catalyzed by (NHC) Pd (II) complexes
GURBUZ N, et al.
Turkish Journal of Chemistry, 39(6), 1115-1157 (2015)
Jang, M.-Y.; De Jonghe, S.; et al.
Tetrahedron Letters, 47, 8917-8917 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service