Skip to Content
Merck
All Photos(1)

Key Documents

221511

Sigma-Aldrich

Sodium 1-butanesulfonate

98%

Synonym(s):

1-Butanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$146.00

$146.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$146.00

About This Item

Linear Formula:
CH3(CH2)3SO3Na
CAS Number:
Molecular Weight:
160.17
Beilstein:
3716662
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$146.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

>300 °C (lit.)

solubility

water: 1 g/10 mL, clear to slightly hazy, colorless

functional group

sulfonic acid

SMILES string

[Na+].CCCCS([O-])(=O)=O

InChI

1S/C4H10O3S.Na/c1-2-3-4-8(5,6)7;/h2-4H2,1H3,(H,5,6,7);/q;+1/p-1

InChI key

XQCHMGAOAWZUPI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 1-butanesulfonate was used in the synthesis of high silicon content SAPO4-5[1]. It was also used as a mobile phase to separate eight selenium compounds (selenite, selenate, selenocystine, selenourea, selenomethionine, selenoethionine, selenocystamine and trimethylselenonium ion)[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue S Han et al.
Chemical communications (Cambridge, England), (2)(2), 166-167 (2002-07-18)
High silicon content SAPO(4)-5 (up to 0.511 atoms per unit cell) has been synthesised, using sodium 3-bromopropanesulfonate, sodium 1-butanesulfonate, sodium naphthalene-1-sulfonate or sodium n-decyl sulfate as surfactants; the SiO2 in the reaction gels ranged up to 3.0 (molar ratio), silicon
J Zheng et al.
Journal of chromatography. A, 874(1), 55-64 (2000-04-18)
For selenium speciation analysis, the hyphenation of chromatographic separation with element-specific detection has proved a useful technique. A powerful separation system, which is capable of resolving several biologically and environmentally important selenium compounds in a single column, is greatly needed.
M T Vincenzini et al.
Life sciences, 31(5), 463-470 (1982-08-02)
The effect of 1-butanesulfonic acid sodium salt and sodium dodecyl sulfate on the activity of highly purified and crystalline enzymes with marked differences in structure and function has been studied. The enzymes were: alcohol dehydrogenase; lactate dehydrogenase; malate dehydrogenase; isocitrate
Tomohiro Narukawa et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(6), 521-527 (2015-06-13)
New measurement conditions for arsenic speciation analysis of rice flour were developed using HPLC-ICP-MS equipped with a reversed phase ODS column. Eight arsenic species, namely, arsenite [As(III)], arsenate [As(V)], monomethylarsonic acid (MMAA), dimethylarsinic acid (DMAA), trimethylarsine oxide (TMAO), tetramethylarsonium (TeMA)
Serena Indelicato et al.
Journal of mass spectrometry : JMS, 48(3), 379-383 (2013-03-16)
The characteristic collision energy (CCE) to obtain 50% fragmentation of positively and negatively single charged noncovalent clusters has been measured. CCE was found to increase linearly with the degrees of freedom (DoF) of the precursor ion, analogously to that observed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service