Skip to Content
Merck
All Photos(1)

Key Documents

220639

Sigma-Aldrich

3-Thiopheneacetic acid

98%

Synonym(s):

3-Thienylacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$146.00
50 G
$428.00

$146.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$146.00
50 G
$428.00

About This Item

Empirical Formula (Hill Notation):
C6H6O2S
CAS Number:
Molecular Weight:
142.18
Beilstein:
113622
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$146.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

98%

mp

73-76 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)Cc1ccsc1

InChI

1S/C6H6O2S/c7-6(8)3-5-1-2-9-4-5/h1-2,4H,3H2,(H,7,8)

InChI key

RCNOGGGBSSVMAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Thiopheneacetic acid acts as monocarboxylate ligand and forms oxo-carboxylate bridged digadolinium(III) complexes[1]. Electrochemical oxidation of 3-thiopheneacetic acid in dry acetonitrile leads to the formation of a conducting polymeric film of poly (3-thiopheneacetic acid)[2].

Application

3-Thiopheneacetic acid was used in:
  • one-step, size control synthesis of gold nanoparticles[3]
  • in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction[4]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Cañadillas-Delgado et al.
Dalton transactions (Cambridge, England : 2003), 39(31), 7286-7293 (2010-07-02)
Two new digadolinium(III) complexes with monocarboxylate ligands, [Gd2(pac)6(H2O)4] (1) and [Gd2(tpac)6(H2O)4] (2) (Hpac = pentanoic acid and Htpac = 3-thiopheneacetic acid), have been prepared and their structures determined by X-ray diffraction on single crystals. Their structures consist of neutral and
One-step, shape control synthesis of gold nanoparticles stabilized by 3-thiopheneacetic acid.
Huang H and Yang X.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 255(1), 11-17 (2005)
Synthesis, density functional theory, molecular dynamics and electrochemical studies of 3-thiopheneacetic acid-capped gold nanoparticles.
Sosibo NM, et al.
Journal of Molecular Structure, 1006(1), 494-501 (2011)
Electrochemistry of poly (3-thiopheneacetic acid) in aqueous solution: evidence for an intramolecular chemical reaction.
Bartlett PN and Dawson DH.
Journal of Materials Chemistry, 4(12), 1805-1810 (1994)
Muhammet Aydın et al.
Macromolecular bioscience, 19(8), e1900109-e1900109 (2019-06-22)
In this study, an impedimetric immunosensor based on polymer poly(thiophene)-graft-poly(methacrylamide) polymer (P(Thi-g-MAm)) modified indium tin oxide (ITO) electrode is developed for the detection of the Neuron Specific Enolase (NSE) cancer biomarker. First, the P(Thi-g-MAm) polymer is synthesized and coated on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service