Skip to Content
Merck
All Photos(2)

Key Documents

220558

Sigma-Aldrich

Ethyl 2-aminothiazole-4-acetate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$92.10

$92.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$92.10

About This Item

Empirical Formula (Hill Notation):
C7H10N2O2S
CAS Number:
Molecular Weight:
186.23
Beilstein:
139617
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$92.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

mp

92-94 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)Cc1csc(N)n1

InChI

1S/C7H10N2O2S/c1-2-11-6(10)3-5-4-12-7(8)9-5/h4H,2-3H2,1H3,(H2,8,9)

InChI key

SHQNGLYXRFCPGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 2-aminothiazole-4-acetate is an organic ligand and possess strong coordination ability and display diverse coordination modes due to the presence of N, O coordination atoms[1].

Application

Ethyl 2-aminothiazole-4-acetate was used in the synthesis of:
  • bis (2-aminothiazole-4-acetato)aquazinc(II)[2]
  • dichloridobis[ethyl 2-(2-amino-1, 3-thiazol-4-yl) acetate-κ2O,N3]cadmium[1]
  • novel heteroaryl-containing benzamide derivatives[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

365.0 °F - closed cup

Flash Point(C)

185 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lai-Jun Zhang et al.
Acta crystallographica. Section E, Structure reports online, 68(Pt 6), m788-m789 (2012-06-22)
The asymmetric unit of the title compound, [CdCl(2)(C(7)H(10)N(2)O(2)S)(2)], contains two complex molecules with similar configurations. The Cd(II) atoms are each six-coordinated by two thiazole N and two carbonyl O atoms from the 2-(2-amino-1,3-thiazol-4-yl)acetate ligand, and by two Cl(-) anions in
Kaapjoo Park et al.
Bioorganic & medicinal chemistry, 22(7), 2280-2293 (2014-03-05)
Novel heteroaryl-containing benzamide derivatives were synthesized and screened using an in vitro assay measuring increases in glucose uptake and glucokinase activity stimulated by 10mM glucose in rat hepatocytes. From a library of synthesized compounds, 3-(4-methanesulfonylphenoxy)-N-[1-(2-methoxy-ethoxymethyl)-1H-pyrazol-3-yl]-5-(3-methyl pyridin-2-yl)-benzamide (19e) was identified as
Lai-Jun Zhang et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 12), m1517-m1517 (2009-01-01)
In the title compound, [Zn(C(5)H(5)N(2)O(2)S)(2)(H(2)O)], the central Zn atom (2 site symmetry) is five-coordinated by two N and three O atoms [Zn-N = 2.047 (3) Å, Zn-O = 2.099 (2) and 1.974 (4) Å] in a distorted square-pyramidal geometry. Besides one O atom from a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service