Skip to Content
Merck
All Photos(1)

Key Documents

219533

Sigma-Aldrich

1-Nitro-1-cyclohexene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO2
CAS Number:
Molecular Weight:
127.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.505 (lit.)

bp

66-68 °C/1.5 mmHg (lit.)

density

1.127 g/mL at 25 °C (lit.)

functional group

nitro

storage temp.

2-8°C

SMILES string

[O-][N+](=O)C1=CCCCC1

InChI

1S/C6H9NO2/c8-7(9)6-4-2-1-3-5-6/h4H,1-3,5H2

InChI key

DJBRXNRKYAWTBL-UHFFFAOYSA-N

General description

Chemoselective hydrogenation of 1-nitro-1-cyclohexene catalyzed by gold nanoparticles supported on TiO2 or Fe2O3 has been reported[1].

Application

1-Nitro-1-cyclohexene was used in the preparation of substituted anilines via chemoselective hydrogenation catalyzed by supported gold nanoparticles (Au/TiO2 and Au/Fe2O3)[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Avelino Corma et al.
Nature protocols, 1(6), 2590-2595 (2007-04-05)
A protocol for the chemoselective hydrogenation of nitro compounds to the corresponding anilines by means of supported gold catalysts is described. Nitro groups on different compounds--containing double bonds, carbonyl, nitrile or amide groups--have been successfully hydrogenated on supported gold nanoparticles
Avelino Corma et al.
Science (New York, N.Y.), 313(5785), 332-334 (2006-07-22)
The selective reduction of a nitro group when other reducible functions are present is a difficult process that often requires stoichiometric amounts of reducing agents or, if H2 is used, the addition of soluble metals. Gold nanoparticles supported on TiO2
Fabian Niedermair et al.
Inorganic chemistry, 49(20), 9333-9342 (2010-09-16)
A series of π-extended phosphorescent palladium(II) and platinum(II) porphyrin complexes were synthesized, in which additional benzene rings are fused radially onto at least one of the four peripheral benzo groups. The photophysical properties of the metalloporphyrins palladium(II)-meso-tetra-(4-fluorophenyl)mononaphthotribenzoporphyrin (Pd1NF), cis-palladium(II)-meso-tetra-(4-fluorophenyl)dibenzodinaphthoporphyrin (Pd2NF)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service