Skip to Content
Merck
All Photos(1)

Key Documents

218332

Sigma-Aldrich

(−)-Limonene oxide, mixture of cis and trans

99%

Synonym(s):

(4S)-1-Methyl-4-(1-methylethenyl)-7-oxabicyclo[4.1.0]heptane, (4S)-1-Methyl-4-(prop-1-en-2-yl)-7-oxabicyclo[4.1.0]heptane

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$339.00

$339.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$339.00

About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$339.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

optical activity

[α]21/D −69°, neat

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.4664 (lit.)

bp

113-114 °C/50 mmHg (lit.)

density

0.929 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CC(=C)[C@H]1CCC2(C)OC2C1

InChI

1S/C10H16O/c1-7(2)8-4-5-10(3)9(6-8)11-10/h8-9H,1,4-6H2,2-3H3/t8-,9?,10?/m0/s1

InChI key

CCEFMUBVSUDRLG-IDKOKCKLSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelli L Hvorecny et al.
Structure (London, England : 1993), 25(5), 697-707 (2017-04-11)
Pseudomonas aeruginosa secretes an epoxide hydrolase with catalytic activity that triggers degradation of the cystic fibrosis transmembrane conductance regulator (CFTR) and perturbs other host defense networks. Targets of this CFTR inhibitory factor (Cif) are largely unknown, but include an epoxy-fatty

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service