Skip to Content
Merck
All Photos(1)

Key Documents

218294

Sigma-Aldrich

(S)-(−)-Perillaldehyde

technical grade, 92%

Synonym(s):

(−)-Perillaaldehyde, (S)-4-Isopropenyl-cyclohexene-1-carboxaldehyde, (S)-p-Mentha-1,8-dien-7-al

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$74.40

$74.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$74.40

About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
2326448
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

$74.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

92%

form

liquid

optical activity

[α]19/D −121°, c = 10 in ethanol

refractive index

n20/D 1.508 (lit.)

bp

104-105 °C/10 mmHg (lit.)

density

0.965 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

CC(=C)[C@H]1CCC(C=O)=CC1

InChI

1S/C10H14O/c1-8(2)10-5-3-9(7-11)4-6-10/h3,7,10H,1,4-6H2,2H3/t10-/m1/s1

InChI key

RUMOYJJNUMEFDD-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service