Skip to Content
Merck
All Photos(1)

Key Documents

217352

Sigma-Aldrich

Methyl 4-chloro-2-nitrobenzoate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)CO2CH3
CAS Number:
Molecular Weight:
215.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Assay

99%

mp

43-45 °C (lit.)

SMILES string

COC(=O)c1ccc(Cl)cc1[N+]([O-])=O

InChI

1S/C8H6ClNO4/c1-14-8(11)6-3-2-5(9)4-7(6)10(12)13/h2-4H,1H3

InChI key

JWOSXVMUUBWGOL-UHFFFAOYSA-N

General description

Methyl 4-chloro-2-nitrobenzoate undergoes fluorodenitration in the presence of tetramethylammonium fluoride to yield methyl 4-chloro-2- fluorobenzoate[1].

Application

Methyl 4-chloro-2-nitrobenzoate was used in the synthesis of methyl 3-hydroxybenzo[b]thiophene-2-carboxylate esters[2].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorodenitrations using tetramethylammonium fluoride.
Boechat N and Clark JH.
Journal of the Chemical Society. Chemical Communications, 11, 921-922 (1993)
Synthesis of Methyl 3-Hydroxybenzo [b] thiophene-2-carboxylate esters by nitro displacement.
Beck JR.
The Journal of Organic Chemistry, 38(23), 4086-4087 (1973)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service