Skip to Content
Merck
All Photos(2)

Key Documents

215988

Sigma-Aldrich

Vanadium(III) oxide

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$137.00
100 G
$522.00

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$137.00
100 G
$522.00

About This Item

Linear Formula:
V2O3
CAS Number:
Molecular Weight:
149.88
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder and chunks

reaction suitability

reagent type: catalyst
core: vanadium

mp

1970 °C (lit.)

density

4.87 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[V]O[V]=O

InChI

1S/3O.2V

InChI key

KFAFTZQGYMGWLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service