Skip to Content
Merck
All Photos(2)

Key Documents

215805

Sigma-Aldrich

Lead(II) titanate

powder, <5 μm, ≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$1,220.00

$1,220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$1,220.00

About This Item

Linear Formula:
PbTiO3
CAS Number:
Molecular Weight:
303.07
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

$1,220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99%

form

powder

particle size

<5 μm

density

7.52 g/mL at 25 °C (lit.)

SMILES string

[PbH2++].[O-][Ti]([O-])=O

InChI

1S/3O.Pb.Ti/q;2*-1;+2;

InChI key

NKZSPGSOXYXWQA-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Synthesis, Properties, and Applications of Perovskite-Phase Metal Oxide Nanostructures

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service