Skip to Content
Merck
All Photos(1)

Documents

215740

Sigma-Aldrich

Disperse Red 1

Dye content 50 %

Synonym(s):

N-Ethyl-N-(2-hydroxyethyl)-4-(4-nitrophenylazo)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N4O3
CAS Number:
Molecular Weight:
314.34
Colour Index Number:
11110
Beilstein:
5353614
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

composition

Dye content, 50%

mp

160-162 °C (lit.)

λmax

503 nm

SMILES string

CCN(CCO)c1ccc(cc1)\N=N\c2ccc(cc2)[N+]([O-])=O

InChI

1S/C16H18N4O3/c1-2-19(11-12-21)15-7-3-13(4-8-15)17-18-14-5-9-16(10-6-14)20(22)23/h3-10,21H,2,11-12H2,1H3/b18-17+

InChI key

FOQABOMYTOFLPZ-ISLYRVAYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

This benchmark dipolar chromophore was incorporated into aromatic polymers to obtain model electrooptic polymer films.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigekatsu Fujii et al.
Journal of peptide science : an official publication of the European Peptide Society, 14(12), 1295-1302 (2008-10-28)
Cyclic octapeptide carrying one or two nonlinear optical chromophores, disperse red 1 (DR-1), was synthesized and immobilized on a substrate to attain an active surface for second-harmonic generation (SHG). Each cyclic octapeptide was transferred on a fused quartz substrate by
Mehmet Cinar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 561-569 (2011-10-01)
This study reports the characterization of disperse red 1 acrylate compound by spectral techniques and quantum chemical calculations. The spectroscopic properties were analyzed by FT-IR, UV-vis, (1)H NMR and (13)C NMR techniques. FT-IR spectrum in solid state was recorded in
Farah Maria Drumond Chequer et al.
Toxicology in vitro : an international journal published in association with BIBRA, 25(8), 2054-2063 (2011-09-13)
Azo dyes constitute the largest class of synthetic dyes. Following oral exposure, these dyes can be reduced to aromatic amines by the intestinal microflora or liver enzymes. This work identified the products formed after oxidation and reduction of the dye
Yaoquan Tu et al.
The journal of physical chemistry. B, 111(14), 3591-3598 (2007-03-29)
We demonstrate a complete procedure for simulations of electric field poled polymeric nonlinear optical systems with the purpose to evaluate the macroscopic electro-optic coefficients. The simulations cover the electric field poling effects on the chromophore order at the liquid state
Roberto Bianchini et al.
Carbohydrate research, 356, 104-109 (2011-12-06)
Commercially available Disperse Orange 29 (1a) and Disperse Red 1 (2a) were elaborated to glycoconjugated species, following a new version of a previously-described 'naturalisation' procedure. Glutamic acid was chosen to achieve a double glycoconjugation, which is essential to give to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service