Skip to Content
Merck
All Photos(2)

Key Documents

215139

Sigma-Aldrich

Pyridine trifluoroacetate

98%

Synonym(s):

Trifluoroacetic acid pyridine salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$234.00

$234.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$234.00

About This Item

Empirical Formula (Hill Notation):
C7H6F3NO2
CAS Number:
Molecular Weight:
193.12
Beilstein:
3735993
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$234.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

chunks

mp

83-86 °C (lit.)

solubility

water: soluble 5%, clear, colorless

functional group

carboxylic acid
fluoro

SMILES string

c1ccncc1.OC(=O)C(F)(F)F

InChI

1S/C5H5N.C2HF3O2/c1-2-4-6-5-3-1;3-2(4,5)1(6)7/h1-5H;(H,6,7)

InChI key

NRTYMEPCRDJMPZ-UHFFFAOYSA-N

General description

Pyridine trifluoroacetate forms reversed micelles with alkylammonium carboxylates in benzene[1].

Application

  • On-tissue chemical oxidation followed by derivatization for mass spectrometry imaging: Investigates a technique for enhancing mass spectrometry imaging of hydroxyl-containing biomolecules using pyridine trifluoroacetate, which could be useful in bioanalytical applications (Wang et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osmotic and activity coefficients of some pyridine complexes with halogenoacetic acids in benzene at 25?C.
Dega-Szapran Z and Szafran M.
Journal of Molecular Liquids, 25(2), 109-113 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service