Skip to Content
Merck
All Photos(1)

Key Documents

214299

Sigma-Aldrich

4-Methyl-2-nitrobenzonitrile

97%

Synonym(s):

2-Nitro-p-tolunitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CH3)CN
CAS Number:
Molecular Weight:
162.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

97-99 °C (lit.)

SMILES string

Cc1ccc(C#N)c(c1)[N+]([O-])=O

InChI

1S/C8H6N2O2/c1-6-2-3-7(5-9)8(4-6)10(11)12/h2-4H,1H3

InChI key

QGBSLPHQCUIZKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methyl-2-nitrobenzonitrile was used in the preparation of 4-cyano-3-nitrobenzyl bromide[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Xu et al.
Biochemistry, 50(39), 8311-8322 (2011-08-24)
Vibrational Stark effect spectroscopy was used to measure electrostatic fields in the hydrophobic region of the active site of human aldose reductase (hALR2). A new nitrile-containing inhibitor was designed and synthesized, and the X-ray structure of its complex, along with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service